Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Kimtech™ G3 Sapphire Nitrile Gloves
BIOPHARMAGREENER_CHOICE

Catalog No. 17004493 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-004-493 Large
17-004-490 X-Small
17-004-491 Small
17-004-492 Medium
17-004-494 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 17-004-493 Supplier Ansell Supplier No. 55878
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nitrile gloves are ideal for medical device, pharmaceutical, or semiconductor manufacturing. Recommended for ISO Class 3 or higher cleanroom environments.

  • Comfortable fit and textured fingertips for a secure grip, high dexterity, and tactile sensitivity
  • Made with nitrile polymer
  • Alternative for workers and technicians with latex allergies
  • Ambidextrous with high tack grip
  • Textured fingertips and beaded cuffs for easy donning
  • AQL 1.5 exceeds ASTM standards for critical defects (pinholes)
  • Double bagged (100 Gloves/Bag, 1000 Gloves/Cs.)
TRUSTED_SUSTAINABILITY

Specifications

Cleanroom Class Class 1 (ISO 3)
Accelerator Free No
AQL 1.5
Color Blue
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 7 mil
Finger Thickness (Metric) 0.18 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.5 mil
Palm Thickness (Metric) 0.14 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size Large
Sterility Non-sterile
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.