Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Kimtech™ G5 Comfort Latex Gloves

Catalog No. 17010047 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-047 X-Large
17-010-044 Small
17-010-045 Medium
17-010-046 Large
17-010-048 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Contains latex. Not appropriate for workers and technicians with latex allergies.
Catalog No. 17-010-047 Supplier Ansell Supplier No. 57711
Add to Cart
Add to Cart
This item is not returnable. View return policy
Contains latex. Not appropriate for workers and technicians with latex allergies.

G5 comfort gloves provide high tactile sensitivity, low residual protein content, and textured palms and fingertips for a secure grip.

  • Beaded cuff to help prevent roll-down
  • Made with natural rubber latex, a renewable resource
  • Ideal for medical device manufacturing, pharmaceuticals, or research labs
  • Recommended for ISO Class 5 or higher cleanroom environments
  • Low residual protein content
  • Easy donning
  • AQL 1.5 exceeds ASTM standards for critical defects (pinholes)
  • Double bagged with case liner
  • 1000 Gloves/Cs. (10 Boxes of 100 Gloves)
TRUSTED_SUSTAINABILITY

Specifications

Cleanroom Class Class 100 (ISO 5)
Accelerator Free No
AQL 1.5
Color Natural
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 7.5 mil
Finger Thickness (Metric) 0.19 mm
Hand Ambidextrous
Latex Free No
Material Latex
Powdered Powder Free
Size X-Large
Sterility Non-sterile
Length (English) 11 in.
Length (Metric) 28 cm
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Recyclable through The RightCycle™ Program.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.