Learn More
Ansell Kimtech™ N95 Pouch Respirator
Catalog No. 17986014 Shop All Ansell Products
Change view
Click to view available options
Size:
One Size
Small
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-986-014 Small
19-005-123 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Not for medical use. Not to be used by healthcare professionals or in healthcare settings.
Catalog No. 17-986-014 Supplier Ansell Supplier No. 54066
Not for medical use. Not to be used by healthcare professionals or in healthcare settings.
N95 pouch respirator is NIOSH-Approved and provides a minimum 95% filtration efficiency of 0.3 micron particles.
  • NIOSH-Approved, provides a minimum 95% filtration efficiency of 0.3 micron particles
  • Guards against the risks posed by airborne particles and contaminants
  • Pouch-style offers the wearer a large breathing chamber to maximize comfort
  • Equipped with two comfort fit headbands and a bendable nosepiece to ensure a consistent seal
  • Not intended for the healthcare market and/or healthcare personnel
  • 300 Kimtech™ N95 Pouch Respirators (6 bags of 50)
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Color White
Design Type Pouch/Duckbill
Disposable Single-use
Filter Type N95
Product Type General Purpose Particulate Respirator
Size Small
With Exhalation Valve No
Filter Efficiency 95%
Nosepiece Type Adjustable Nose Clip
Sterility Non-sterile
Strap Attachment Type Two Comfort Fit Headbands
FDA Cleared Surgical Mask No