Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell Kimtech™ Polaris™ Nitrile Exam Gloves
GREENER_CHOICE

Catalog No. 17103196A Shop All Ansell Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-103-196A X-Small
17-103-196B Small
17-103-196C Medium
17-103-196D Large
17-103-196E X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-103-196A Supplier Ansell Supplier No. 62770
Add to Cart
Add to Cart
This item is not returnable. View return policy

Kimtech™ is your partner in driving progress. Tested against some of the harshest chemicals used in your work environment, Kimtech™ Polaris™ Exam Nitrile Gloves meet and exceed the industry standards for exam gloves.

  • The proprietary high-quality nitrile formulation delivers the highest level of protection in the Kimtech™ exam glove portfolio and has been tested for use with chemotherapy drugs and fentanyl.
  • With an average tensile strength that is more than double the industry standard, Kimtech™ Polaris™ provides the strength and comfort needed for performance and productivity; it's even certified by US Ergonomics.
  • Eligible for recycling through The RightCycle™ Program, 100% of your non-hazardous Kimtech™ Polaris™ Nitrile Exam Gloves can be diverted from the landfill.
  • With protection, durability, and comfort you can count on, these gloves are built for science, made for you
  • Ambidextrous
TRUSTED_SUSTAINABILITY

Specifications

Chlorinated No
Accelerator Free No
Applications Laboratory
AQL 0.65
Color Dark Magenta
Disposable Single-use
Exterior Finish Fingertip Textured
Finger Thickness (English) 5.91 mil
Finger Thickness (Metric) 0.15 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 5.91 mil
Palm Thickness (Metric) 0.15 mm
Powdered Powder Free
Product Type Nitrile Examination Glove
Size X-Small
Sterility Non-sterile
Thickness (English) 5.91 mil
Thickness (Metric) 0.15 mm
FDA 510 (k) Yes
Glove Cuff Style Beaded
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.