Learn More
Ansell Kimtech™ A7 Ankle High Shoe Covers
BIOPHARMAGREENER_CHOICE
Catalog No. 19169981 Shop All Ansell Products
Change view
Click to view available options
Size:
Universal
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-169-981 X-Large
19-169-980 Universal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-169-981 Supplier Ansell Supplier No. 47973
Add to Cart
Add to Cart
This item is not returnable. View return policy
Ankle high shoe covers provide extra contamination protection by covering the shoelace area with two elastic bands--one above the ankle and one below the ankle.
  • Ankle high shoe covers provide 30% more protection that a shoe cover, but less than a boot
  • Two elastic bands placed both above and below the ankle for enhanced fit and coverage
  • Comfortably cover the shoelace area, closing a gap that could be a vehicle for contamination
  • Deliver a higher barrier of liquid protection and a securer grip with anti-skid vinyl soles
  • Kimtech A7 material offers enhanced chemical and biological protection (passes ASTM F1670 for penetration of blood and body fluids)
  • Recyclable through the RightCycle Program, which turns them into the raw materials to create eco-friendly consumer goods
  • Packaged 300 Shoe Covers/Case, 3 Bags of 100 Covers
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Material SMS
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Over and Under Elastic Ankle Bands
Sole Type Anti-skid
Certifications/Compliance ASTM F1670
Packaging Quantity 300/Cs.
Packaging Type 3 Bags of 100 Covers
Product Type Controlled Environments Ankle High Shoe Covers
Show More Show Less
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.