Learn More
Cancer Diagnostics, Inc.™ Kimtech Pure™ M5 Face Masks
SDP
Catalog No. 22500458 Shop All Cancer Diagnostics Products
Change view
Click to view available options
Includes:
Faceshield Only, 100/Cs.
Visors with Clear Nosepieces, 10/Pk.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
22-500-458 Visors with Clear Nosepieces, 10/Pk.
22-500-456 -
22-500-457 Faceshield Only, 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 22-500-458 Supplier Cancer Diagnostics, Inc.™ Supplier No. CLPF10
Add to Cart
Add to Cart
This item is not returnable. View return policy
Lightweight and comfortable for extended work sessions
  • Reusable, economical, and disposable protection weighs less than 1 1/2 oz.
  • Visor is supported by a clear nosepiece; remove the nosepiece and it fits over most prescription eyewear
  • Eliminates glare from overhead lighting
Order Info
Available in LabPak with three visors (in assorted colors), clear nosepieces, and 15 disposable clear shields; replacement visor and shield also sold separately.
Specifications
Product Type Replacement Faceshield Visor
For Use With (Equipment) ClearLITE Plus
Frame Color Assorted
Includes Visors with Clear Nosepieces, 10/Pk.
No. per Pack 10/Pk.