Learn More
Alkali Scientific Kimtech Science Kimwipes (34120) Delicate Task Wipers, 1-Ply, 4.4" x 8.4" (11.18 x 21.34 cm), 286/Box
SDP
Supplier:  Alkali Scientific KM066
Encompass_Preferred
Kimwipes are the best choice for cleaning laboratory surfaces, sensitive instruments, laboratory lenses, and health care settings. They easily clean liquids, dust, and small particles, and the anti-static dispensing design reduces electrostatic discharge. They are 1-ply, low-lint, and are great for cleaning any research lab. White, 286 sheets per box. Pop-up dispenser box. 4" x 8.4". For general, low-lint cleaning tasks.
Catalog No. 50-238-9555
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.