Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Kinase buffer II (5X)

Catalog No. p-5394072
Change view
Click to view available options
Quantity:
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60269AE 100 mL
AAJ60269AK 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60269AE Supplier Thermo Scientific™ Supplier No. J60269AE
Only null left
Add to Cart

For cellular and molecular biology applications

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Formulation: 100mM MOPS, 50mM magnesium acetate, 5mM DTT 5, 5mM EDTA, 25mM beta glycerophosphate, and 5mM sodium orthovanadate, pH 7.4
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kinase buffer II (5X)
Physical Form Liquid
Quantity 100 mL
Formulation 100mM MOPS, 50mM magnesium acetate, 5mM DTT 5, 5mM EDTA, 25mM beta glycerophosphate, and 5mM sodium orthovanadate, pH 7.4

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.