Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alexa Fluor™ Kinase Tracer 1710, 25 μM

Catalog No. 502576143
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-257-6143 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-257-6143 Supplier Alexa Fluor™ Supplier No. PV6088
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Alexa Fluor™ Kinase Tracer 1710 is detected by addition of a Eu-labeled anti-tag antibody.

  • Excitation/Emission (nm): 340/615, 665
  • Label or Dye: Alexa Fluor™ 647
  • Target Enzyme: Kinases (Multiple)
  • Validated Kinases: PIK3CA, ABL1, ABL1 E255K, ABL1 G250E, ABL1 Y253F, WNK3
  • In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 1710 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps.
TRUSTED_SUSTAINABILITY

Specifications

Assay Kinase Binding Assay
Assay Entry Biochemical binding/kinetic
Content And Storage Store at -20°C.
Excitation/Emission 340/615, 665
For Use With (Application) TR-FRET
For Use With (Equipment) Microplate Reader
Format 384-well Plate
Label or Dye Alexa Fluor™ 647
Quantity 25 μL
Readout Kinetic
Shipping Condition Dry Ice
Target Entry ABL1 G250E, ABL1 E255K, ABL1 Y253F, ABL1, PIK3CA/PIK3R1, WNK3
Type Kinase Tracer
Validated Kinases PIK3CA, ABL1, ABL1 E255K, ABL1 G250E, ABL1 Y253F, WNK3
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.