Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Kinase Tracer 178, 25 μM

Catalog No. PV5593
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5593 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5593 Supplier Thermo Scientific™ Supplier No. PV5593
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 178 to a kinase is detected by addition of a Eu-labeled anti-tag antibody.

In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 178 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps.

TRUSTED_SUSTAINABILITY

Specifications

Product Line LanthaScreen
For Use With (Equipment) Microplate Reader
Target Kinases (Multiple)
For Use With (Application) TR-FRET
Assay Kinase Binding Assay
Content And Storage Store at -20°C.
Assay Entry Biochemical binding/kinetic
Label or Dye Alexa Fluor 647
Product Type Kinase Tracer
Quantity 25 μL
Format 384-well plate
Readout Kinetic
Shipping Condition Dry Ice
Target Entry PTK6, DDR2, BMPR1A, RAF1 (cRAF) Y340D Y341D, ACVR1B, WNK2, TEC, DDR1, EPHB4, ERBB4, KIT V654A, TGFBR1, NLK, TGFBR2, EPHB3, BRAF V599E, BRAF, ACVR2B, ACVR2A
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.