Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Kinase Tracer 236, 50 μM

Catalog No. PV5592
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PV5592 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PV5592 Supplier Thermo Scientific™ Supplier No. PV5592
  • $670.00
    $622.65 / Each of 1

    Save $47.35

    7% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 236 to a kinase is detected by addition of a Eu-labeled anti-tag antibody.

In the LanthaScreen™ Eu Kinase Binding Assay, binding of an Alexa Fluor™ conjugate or tracer such as Kinase Tracer 236 to a kinase is detected by addition of a Eu-labeled anti-tag antibody. Binding of the tracer & antibody to a kinase results in a high degree of FRET, whereas displacement of the tracer with a kinase inhibitor results in a loss of FRET. Unlike many kinase activity assays, this assay is a simple mix and read assay, with no development steps.

TRUSTED_SUSTAINABILITY

Specifications

Assay Kinase Binding Assay
Assay Entry Biochemical binding/kinetic
Content And Storage Store at -20°C.
For Use With (Application) TR-FRET
For Use With (Equipment) Microplate Reader
Format 384-well Plate
Label or Dye Alexa Fluor™ 647
Quantity 25 μL
Readout Kinetic
Shipping Condition Dry Ice
Target Entry CSF1R, CSK, EPHA5, PDGFRA T674I, PDGFRA V561D, RET Y791F, SYK, TYRO3, ZAP70, MAP4K5, PRKCQ, PRKX, ROCK2, IKBKE, PAK4, MAP3K2, LRRK2 D1994A, GRK7, GSK3A, GSK3B, SNF1LK2, MAP2K1 S218D S222D, FGFR1, FGFR2, MAP3K10, MAP3K9, JAK2 JH1 JH2, AMPK (A2/B1/G1), CAMK2B, CHEK1, PIM2, PRKCN, PRKD2, PRKG2, RPS6KA5, MELK, BLK, EPHB2, IGF1R, LYNA, MERTK, RET V804L, ROS1, TEK, TTK, SGK, CHUK, IKBKB, TAOK3, TYK2, IRAK1, MYLK2, SRPK2, STK4, FLT1, MUSK, CAMK2D, LRRK2 R1441C, DYRK1A, DYRK3, MAP2K1, MAP2K6, RPS6KA6, BMX, EPHA4, FYN, PTK2, PTK2B, RET, MAP4K2, MINK1, GRK6, JAK3, LRRK2, AMPK (A1/B1/G1), AURKA, CAMK1D, CAMK4, PIM1, PKN1, PRKD1, PRKG1, RPS6KA4, STK22D, NUAK1, EPHA3, FLT3, FLT4, INSR, LCK, YES1, KDR, MET, TNK2, PRKCI, CAMK2A, RPS6KA3, MAP3K3, ABL2, AXL, FGFR3, FGFR3 K650E, FGR, HCK, AURKB, TAOK2, MAP3K11, CAMKK1, CDC42BPA, CDK1, CDK5, CDK8, CDK9/Cyclin K, MAP2K3, CSNK1G3, DMPK, DYRK1B, ACVR1, EPHA1, EPHA8, LYNB, MST1R, PDGFRA D842V, MAP4K4, LRRK2 G2019S, CAMKK2, CDC42BPB, CDK2, CDK7, CDK9/Cyclin T1, EPHA2, FER, LTK, PDGFRA, PDGFRB, MARK4, RPS6KB1, EGFR T790M, EPHA7, EPHB1, PHKG1, ROCK1, RPS6KA1, TBK1, JAK2, MARK1, PHKG2, MLCK, PAK6, SLK, LIMK1, MKNK2, MYLK, FRK, ITK, LRRK2 I2020T, BRSK1, CLK2, PDK1, MAP2K2, MAP3K14, MAP2K6 S207E T211E, FLT3 D835Y, SRC, GRK5, GRK4, STK3, BTK, FES, KIT T670I, RPS6KA2, SGKL, STK25, PAK7, PRKACA, CLK1, CLK4, MARK2, STK17A, PRKCH, EIF2AK2, MAP3K5, LIMK2, STK16
Type Kinase Tracer
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.