Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kindlin-2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501568091
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-8091 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-8091 Supplier Bethyl Laboratories, Inc Supplier No. A304577AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

FERMT2 participates in the connection between ECM adhesion sites and the actin cytoskeleton and also in the orchestration of actin assembly and cell shape modulation. It recruits migfilin (FBLP1) protein to cell-ECM focal adhesion sites.

Specifications

Antigen Kindlin-2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene FERMT2
Gene Accession No. Q8CIB5, Q96AC1
Gene Alias KIND2, MIG2, PLEKHC1, UNC112, UNC112B, mig-2, PH domain-containing family C member 1, fermitin family homolog 2, kindlin 2, kindlin-2, mitogen inducible gene 2 protein, mitogen-inducible gene 2 protein, pleckstrin homology domain containing, family C (with FERM domain) member 1, pleckstrin homology domain c
Gene Symbols FERMT2
Host Species Rabbit
Immunogen Between 175 and 225
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10979, 218952
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.