Learn More
Specifications
Specifications
| Gene ID (Entrez) | 3798 |
| Species | Human |
| Product Line | Novus |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-1058 |
| Gene Alias | D12S1889, KIF5A variant protein, kinesin family member 5 A, kinesin heavy chain isoform 5 A, Kinesin heavy chain neuron-specific 1, kinesin, heavy chain, neuron-specific, MY050, Neuronal kinesin heavy chain, NKHC1, NKHCspastic paraplegia 10 (autosomal dominant), SPG10 |
| Gene Symbol | KIF5A |
| Quantity | 0.1 mg |
| Research Category | Cell Biology, Cellular Markers |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
