Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kinesin 5A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1642 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1642 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1642 Supplier Invitrogen™ Supplier No. PA1642
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-642 detects kinesin 5A from mouse, human, and bovine samples. This antibody is specific for kinesin 5A and does not detect other kinesin isotypes. PA1-642 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~110 kDa protein representing kinesin 5A from mouse neuronal cell lysate and human retinal extracts. The PA1-642 immunizing peptide corresponds to amino acid residues 1007-1027 from mouse kinesin 5A. This sequence is completely conserved in human. This peptide (Cat. # PEP-197) is available for use in neutralization and control experiments.

This gene encodes a member of the kinesin family of proteins. Members of this family are part of a multisubunit complex that functions as a microtubule motor in intracellular organelle transport. Mutations in this gene cause autosomal dominant spastic paraplegia 10.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kinesin 5A
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide
Gene KIF5A
Gene Accession No. P33175, Q12840
Gene Alias D10Bwg0738e; D12S1889; Khc; Kiaa4086; Kif5; Kif5a; kif5a {ECO:0000312; KIF5A variant protein; kinesin family member 5A; kinesin heavy chain isoform 5A; kinesin heavy chain isoform 5A; kinesin heavy chain; kinesin heavy chain neuron-specific 1; kinesin, heavy chain, neuron-specific; Kns; mKIAA4086; MY050; Neuronal kinesin heavy chain; NKHC; Nkhc1; RGD:1303035}; SPG10
Gene Symbols KIF5A
Host Species Rabbit
Immunogen Synthetic Peptide: C(1007) G Y E A E D Q A K L F P L H Q E T A A S(1027).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16572, 3798, 511273
Target Species Human, Mouse, Bovine
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.