Learn More
Description
PA1-643 detects kinesin 5B from mouse and human samples. This antibody is specific for kinesin 5B and does not detect other kinesin isotypes. PA1-643 has been successfully used in Western blot and immunofluorescent procedures. By Western blot, this antibody detects an ~110 kDa protein representing kinesin 5B protein. The PA1-643 immunizing peptide corresponds to amino acid residues 376-396 from human kinesin 5B. This sequence is 95% and 90% conserved for mouse and rat, respectively. This peptide (Cat. # PEP-198) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | Kinesin 5B |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1 mg/mL BSA and 0.05% sodium azide |
| Gene | KIF5B |
| Gene Accession No. | P33176, Q2PQA9, Q61768 |
| Gene Alias | AL022807; Conventional kinesin heavy chain; EMBL:ABC25059.1}; epididymis secretory protein Li 61; HEL-S-61; Khc; Khcs; KIF5A; KIF5B; kif5b {ECO:0000312; kinesin 1 (110-120kD); kinesin family member 5A; kinesin family member 5B; kinesin heavy chain; kinesin heavy chain member 5B, ubiquitous; kinesin-1 heavy chain; KINH; KNS; Kns1; ubiquitous kinesin heavy chain; UKHC; U-KHC; Unknown (protein for IMAGE:7944745) |
| Gene Symbols | KIF5B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
