Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kinesin 5C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA1644 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA1644 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA1644 Supplier Invitrogen™ Supplier No. PA1644
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA1-644 detects kinesin 5C from human samples. This antibody is specific for kinesin 5C and does not detect other kinesin isotypes. No cross-reactivity is observed using bovine or mouse samples. PA1-644 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~110 kDa protein representing kinesin 5C from human retinal extract. An unidentified band is also detected at ~100 kDa. The PA1-644 immunizing peptide corresponds to amino acid residues 938-957 from human kinesin 5C. This peptide (Cat. # PEP-199) is available for use in neutralization and control experiments.

Kinesins are a superfamily of microtubule-associated motor proteins involved in a variety of cellular processes including membranous organelle transport and cell division. Kinesin has been found in a variety of organisms and cell types and is subject to spatial and temporal regulation. These proteins have a modular structure including a conserved motor domain of approximately 350 amino acids, which is responsible for microtubule binding and ATP hydrolysis. In addition to the motor domain, subfamily members share common domain organization, exhibit sequence similarity, motility properties, and cellular functions outside of the motor domain. There are currently three known Kinesin 5 family members denoted as A, B, and C. Kinesin 5A and kinesin 5C appear to be exclusively neuronal, whereas kinesin 5B appears to be ubiquitous in its expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kinesin 5C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1 mg/mL BSA and 0.05% sodium azide
Gene KIF5C
Gene Accession No. O60282
Gene Alias CDCBM2; Khc; KIAA0531; KIF5C; kinesin family member 5C; kinesin heavy chain isoform 5C; kinesin heavy chain member 5C; kinesin heavy chain neuron-specific 2; KINN; NKHC; NKHC2; NKHC-2
Gene Symbols KIF5C
Host Species Rabbit
Immunogen Synthetic Peptide: C A(938) V H A I R G G G G S S S N S T H Y Q K(957).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3800
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.