Learn More
Cole-Parmer Heraeus Noblelight Hollow Cathode Lamp, palladium, 50 mm, Perkin Elmer Lumina coded
Supplier:  Cole-Parmer 1195171
Encompass
Kinesis palladium (Pd) single-element hollow cathode lamps are quick to reach equilibrium performance, offer a long-term constant light intensity, and low noise with good calibration. Inside each lamp, there is a high volume of inert gas to increase the lamps lifetime to 5000 mA hours. Choose from coded lamp versions for Perkin Elmer, Varian/Agilent, and Unicam systems.
Catalog No. NC2186915
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.