Learn More
Description
- GL45 thread bottles suitable for suitable for either vacuum or positive pressure applications
- Indelible volumetric markings
- Chemically-resistant borosilicate glass
Specifications
Specifications
| Capacity (Metric) | 1000 mL |
| Color | Clear |
| Material | Borosilicate Glass |
| Capacity (English) | 32 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.