Learn More
Medchemexpress LLC Adenosine, N-(2-furanylmethyl)- | 4338-47-0 | MFCD00037987 | 99.9% | 347.33 g/mol | C15H17N5O5 | 50 MG

Supplier: Medchemexpress LLC HY10105550MG
Kinetin riboside (Adenosine, N-(2-furanylmethyl)-) is a ribosylated cytokinin analog used in biochemical and cell biology research. It induces apoptosis in cancer cells and inhibits proliferation of HCT-15 cells (IC50 = 2.5 μM). The reagent is supplied as a high-purity solid and is intended for cell-based assays investigating apoptosis, proliferation, and cytotoxicity.
- High purity: 99.88%.
- Molecular weight: 347.33 g/mol.
- Chemical formula: C15H17N5O5.
- Appearance: solid, white to off-white.
- Storage: powder -20°C for long term; in solvent -80°C recommended.
- Reported bioactivity: induces apoptosis and inhibits HCT-15 cell proliferation (IC50 2.5 μM).
- Pack sizes suitable for small-scale research, including 50 mg.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.