Learn More
Thermo Scientific™ KingFisher 96 Deep-well Plates
Description
- KingFisher™ Plastics for Automated DNA, RNA, and Protein Purification
- KingFisher™ plastics consumables—including plates, strips, and tip combs—are specifically designed for automated magnetic bead-based purification of DNA, RNA, proteins, and cells. Engineered for easy compatibility with KingFisher Apex™, Flex™, Duo Prime, or Presto™ instruments, they enable reliable, high-throughput sample processing in research and clinical laboratories.
- Purpose-Built for Magnetic Bead Workflows
- Unlike standard lab plastics, KingFisher consumables are optimized for magnetic particle processing. Carefully designed plate formats, well geometries, and tip comb structures helps ensure efficient magnetic bead capture, transfer, and mixing—key factors that directly impact yield, purity, and reproducibility.
- Tip combs protect magnetic rods while enabling precise bead handling, and plate designs support optimal reagent interaction throughout the purification workflow
- Consistent Quality Through Controlled Manufacturing
- Manufactured using high-purity, traceable raw materials in a fully automated, quality-controlled process, KingFisher plastics help deliver consistent performance across runs. This enables reliability in demanding applications where reproducibility is critical.
Each consumable is engineered to limit variability and support standardized workflows across laboratories.
Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex, KingFisher Duo Prime and KingFisher Presto.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
| Barcode | Unique Barcode |
| Content And Storage | KingFisher 96 Deep-Well Plate, Unique Barcode, KingFisher Apex |
| Product Type | Deep-well Plate |
| For Use With (Equipment) | KingFisher Apex |
| No. of Wells | 96 |
| Quantity | 50/Cs. |
For research use only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.