Learn More
Thermo Scientific™ KingFisher™ Flex™ Purification System Accessories

Description
Find a flexible solution for different types of sample processing needs. With 24 magnet heads, customers can raise the processing volume up to 5 mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.
- Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
- High throughput system processes up to 96 samples in less than 20 minutes.
- Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands.
- Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification.
- Ready-made protocols for different type of applications are available.
Recommended for:
- Genomics and proteomics
- Target identification
- Veterinary assays
- Biomarker discovery
- Quality control
- Forensics
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Product Type | Flex 96 DW Head, heating block |
| For Use With (Equipment) | KingFisher™ Flex™ Purification System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.