Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ KingFisher™ Flex™ Purification System Accessories
GSA_VA

Catalog No. 22387012
Change view
Click to view available options
Product Type:
Flex 24 DW head, heating block
Flex 96 DW Head, heating block
Flex 96 KF Head, heating block
Flex 96 PCR Head
Flex 96 PCR Head, heat block
Flex 96 PCR heating block
Flex KF Head
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
22-387-012 Flex 24 DW head, heating block
22-387-009 Flex 96 PCR Head, heat block
22-387-017 Flex 96 PCR heating block
22-387-014 Flex KF Head
22-387-013 Flex 96 PCR Head
22-387-010 Flex 96 KF Head, heating block
22-387-011 Flex 96 DW Head, heating block
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 22-387-012 Supplier Thermo Scientific™ Supplier No. 24074441
Add to Cart
Add to Cart
Request a Quote

Prepare samples efficiently from a variety of materials with Thermo Scientific™ KingFisher™ Flex Purification Systems which offer highly versatile, automated magnetic-particle processing for DNA/RNA, protein, or cell purification from virtually any source.

Find a flexible solution for different types of sample processing needs. With 24 magnet heads, customers can raise the processing volume up to 5 mL, resulting in increased yield of the purified product. For higher throughput needs, 96 samples can be processed with 96 magnet head.

  • Fully automated system yields high-speed purification of nucleic acids, proteins and cells.
  • High throughput system processes up to 96 samples in less than 20 minutes.
  • Open and flexible system lets the customer use any magnetic particle-based kit to meet the application demands.
  • Easy-to-use Thermo Scientific™ BindIt™ Software provides instrument control, protocol creation and modification.
  • Ready-made protocols for different type of applications are available.

Recommended for:

  • Genomics and proteomics
  • Target identification
  • Veterinary assays
  • Biomarker discovery
  • Quality control
  • Forensics

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Flex 24 DW head, heating block
For Use With (Equipment) KingFisher™ Flex™ Purification System
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.