Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ KingFisher™ Plastics for 96 standard and PCR formats

Catalog No. 14387355
Change view
Click to view available options
For Use With (Equipment):
KingFisher Apex
KingFisher Flex
KingFisher Flex and Presto
Quantity:
100/Cs.
48/Cs.
50/Cs.
80/Cs.
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Quantity
14-387-355 KingFisher Apex 48/Cs.
97-002-560 KingFisher Apex 50/Cs.
22-387-030 KingFisher Flex and Presto 48/Cs.
22-387-028 KingFisher Flex 100/Cs.
22-387-027 KingFisher Flex 80/Cs.
14-387-353 KingFisher Apex 100/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-387-355 Supplier Thermo Scientific™ Supplier No. 97002540B
Add to Cart
Add to Cart

Optimize your performance using Thermo Scientific™ KingFisher consumables designed specifically for use with the KingFisher Flex, Duo Prime, and Presto instruments.

Optimize your performance using Thermo Scientific™ KingFisher Plastics designed specifically for use with the KingFisher Flex, Duo Prime, Presto and Apex instruments. KingFisher instruments utilize disposable plastic plates and tip combs made of polypropylene, which are ideal for magnetic particle processing due to their low binding affinity for biomolecules. Both the yield and quality of the isolated protein or nucleic acid are significantly improved with special plates and tip combs designed for KingFisher systems.

Use With: Thermo Scientific™ KingFisher instruments, including KingFisher Apex, KingFisher Flex and KingFisher Presto.
TRUSTED_SUSTAINABILITY

Specifications

Capacity 200 μL
Content And Storage KingFisher 96 Plate (200 μL), Barcoded
For Use With (Equipment) KingFisher Apex
Quantity 48/Cs.

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.