Learn More
Thermo Scientific™ KingFisher™ Systems Consumables
Description
Optimize performance using Thermo Scientific™ KingFisher™ Systems Consumables, specifically designed for use with the KingFisher System and ideal for magnetic particle processing because of their low binding affinity for biomolecules. The specially-designed, disposable tip combs and microplates, which are made of polypropylene, enable excellent recovery of magnetic beads.
- Specifically designed for use with the KingFisher System
- Ideal for magnetic particle processing because of their low binding affinity for biomolecules
- The specially-designed, disposable tip combs and microplates are made of polypropylene
- This enables excellent recovery of magnetic beads
Specifications
Specifications
| Capacity | 100 μL |
| For Use With (Application) | Magnetic particle processing |
| For Use With (Equipment) | KingFisher System |
| Includes | KingFisher plate 100 μL |
| Quantity | 50/Cs. |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.