Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kininogen Heavy Chain Monoclonal Antibody (2B5)
GREENER_CHOICE

Catalog No. 01671860
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-671-860 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-671-860 Supplier Invitrogen™ Supplier No. MA110832
  • $872.00
    $697.00 / Each of 1

    Save $175.00

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-10832 detects High MW Kininogen, heavy chain, from human samples. MA1-10832 has been successfully used in ELISA and Western blot applications. The MA1-10832 immunogen is purified high molecular weight kininogen.

This gene uses alternative splicing to generate two different proteins- high molecular weight kininogen (HMWK) and low molecular weight kininogen (LMWK). HMWK is essential for blood coagulation and assembly of the kallikrein-kinin system. Also, bradykinin, a peptide causing numerous physiological effects, is released from HMWK. In contrast to HMWK, LMWK is not involved in blood coagulation. Three transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kininogen Heavy Chain
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene KNG1
Gene Accession No. P01042
Gene Alias Alpha-2-thiol proteinase inhibitor; BDK; BK; Bradykinin; fitzgerald factor; High molecular weight kininogen; HMWK; Ile-Ser-Bradykinin; Kallidin I; Kallidin II; kininogen 1; kininogen-1; Kininogen-1 heavy chain; Kininogen-1 light chain; KNG; KNG1; Low molecular weight growth-promoting factor; Lysyl-bradykinin; T-kinin; Williams-Fitzgerald-Flaujeac factor
Gene Symbols KNG1
Host Species Mouse
Immunogen Purified high molecular weight kininogen.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3827
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.