Learn More
Description
Specifications
Specifications
| For Use With (Application) | Antibody Production |
| Gene ID (Entrez) | 57292 |
| Name | killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A |
| Quantity | 10 μg |
| Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Gene Alias | CD158F, KIR2DL5, KIR2DL5.1, KIR2DL5.3 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.