Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KIR2DL5A/KIR2DL5B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568460
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568460 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568460 Supplier Invitrogen™ Supplier No. PA568460
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with Human samples.

KIR2DL5A and KIR2DL5B are Killer cell immunoglobulin-like receptors (KIRs), transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. The KIR genes are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC). The gene content of the KIR gene cluster varies among haplotypes, although several 'framework' genes are found in all haplotypes. The KIR proteins are classified by the number of extracellular immunoglobulin domains (2D or 3D) and by whether they have a long (L) or short (S) cytoplasmic domain. KIR proteins with the long cytoplasmic domain transduce inhibitory signals upon ligand binding via an immune tyrosine-based inhibitory motif (ITIM), while KIR proteins with the short cytoplasmic domain lack the ITIM motif and instead associate with the TYRO protein tyrosine kinase binding protein to transduce activating signals. KIR proteins are thought to play an important role in regulation of the immune response. Functionally, KIR2DL5A and KIR2DL5B are receptors on natural killer (NK) cells for HLA-C alleles and prevent cell lysis by inhibiting the activity of NK cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KIR2DL5A/KIR2DL5B
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene KIR2DL5A
Gene Accession No. Q8N109, Q8NHK3
Gene Alias CD158 antigen-like family member F2; CD158F; CD158F1; CYB561; CYB561A1; Cytochrome b561; cytochrome b-561; cytochrome b561 family, member A1; ferric-chelate reductase 2; FRRS2; killer cell Ig-like receptor; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 5A; killer cell immunoglobulin like receptor, two Ig domains and long cytoplasmic tail 5B; killer cell immunoglobulin-like receptor 2DL5A; killer cell immunoglobulin-like receptor 2DL5B; killer cell immunoglobulin-like receptor 2DLX; killer cell immunoglobulin-like receptor KIR2DL5A; killer cell immunoglobulin-like receptor two domains long cytoplasmic tail 5; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A; killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5B; killer Ig receptor; killer-cell Ig-like receptor; KIR2DL5; KIR2DL5.1; KIR2DL5.2; KIR2DL5.3; KIR2DL5A; KIR2DL5B; KIR2DLX
Gene Symbols KIR2DL5A, KIR2DL5B
Host Species Rabbit
Immunogen Synthesized peptide derived from human KIR2DL5A/KIR2DL5B [aa 121-170].
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 553128, 57292
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.