Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kir6.1 (KCNJ8) Monoclonal Antibody (N366/60)

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527679
Description
A 1:100 dilution of MA5-27679 was sufficient for detection of Kir6.1 in 20 μg of mouse brain lysate by ECL immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 45kDa. This antibody was formerly sold as clone S366-60.
Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. KCNJ8 is an integral membrane protein and inward-rectifier type potassium channel. KCNJ8, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins.Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins.
Specifications
| Kir6.1 (KCNJ8) | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.4 | |
| P97794, Q15842, Q63664 | |
| KCNJ8 | |
| Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (PFA fixed), Western Blot, Immunocytochemistry | |
| N366/60 | |
| Unconjugated | |
| KCNJ8 | |
| AI448900; ATP-sensitive inward rectifier potassium channel 8; ATP-sensitive inward rectifier potassium channel Kir6.1; gnite; Inward rectifier K(+) channel Kir6.1; inward rectifier potassium channel 6.1; inward rectifier potassium channel Kir6.1; inwardly rectifier K(+) channel Kir6.1; Inwardly rectifying potassium channel gene subfamily J-8 (ATP sensitive); Inwardly rectifying potassium channel gene, subfamily J-8 (ATP sensitive); inwardly rectifying potassium channel Kir6.1; ion channel; Kcnj8; KCNJ8 protein; Kir6.1; Kir6.1 like; potassium channel, inwardly rectifying subfamily J member 8; potassium channel, inwardly rectifying subfamily J, member 8; potassium inwardly rectifying channel subfamily J member 8; potassium inwardly-rectifying channel J8; potassium inwardly-rectifying channel, subfamily J, member 8; potassium voltage-gated channel subfamily J member 8; si:dkey-183c2.2; slmbr; sltr; UKATP1; uKATP-1; zkir6.1 | |
| Mouse | |
| Protein G | |
| RUO | |
| 16523, 25472, 3764 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction