Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Kir6.1 (KCNJ8) Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556628
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556628 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556628 Supplier Invitrogen™ Supplier No. PA556628
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KRSPLYDISA TDLANQDLEV IVILEGVVET TGITTQARTS YIAEEIQWGH RFVSIVTEEE GVYSVDYSKF GNTVKVAAPR CSARELDEKP SILIQTLQKS ELSHQNSLRK RNSMRRNNSM RRNNSIRRNN SSLMVPKVQF MTPEGNQNT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.

Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. KCNJ8 is an integral membrane protein and inward-rectifier type potassium channel. KCNJ8, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins.Potassium channels are present in most mammalian cells, where they participate in a wide range of physiologic responses. The protein encoded by this gene is an integral membrane protein and inward-rectifier type potassium channel. The encoded protein, which has a greater tendency to allow potassium to flow into a cell rather than out of a cell, is controlled by G-proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Kir6.1 (KCNJ8)
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNJ8
Gene Accession No. Q15842
Gene Alias AI448900; ATP-sensitive inward rectifier potassium channel 8; ATP-sensitive inward rectifier potassium channel Kir6.1; gnite; Inward rectifier K(+) channel Kir6.1; inward rectifier potassium channel 6.1; inward rectifier potassium channel Kir6.1; inwardly rectifier K(+) channel Kir6.1; Inwardly rectifying potassium channel gene subfamily J-8 (ATP sensitive); Inwardly rectifying potassium channel gene, subfamily J-8 (ATP sensitive); inwardly rectifying potassium channel Kir6.1; ion channel; Kcnj8; KCNJ8 protein; Kir6.1; Kir6.1 like; potassium channel, inwardly rectifying subfamily J member 8; potassium channel, inwardly rectifying subfamily J, member 8; potassium inwardly rectifying channel subfamily J member 8; potassium inwardly-rectifying channel J8; potassium inwardly-rectifying channel, subfamily J, member 8; potassium voltage-gated channel subfamily J member 8; si:dkey-183c2.2; slmbr; sltr; UKATP1; uKATP-1; zkir6.1
Gene Symbols KCNJ8
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNJ8. Recombinant protein control fragment (Product #RP-101008).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3764
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.