Learn More
AdipoGen Kirromycin
SDP
Supplier:  AdipoGen BVT0157M005
Encompass_Preferred
Chemical. CAS 50935-71-2. Formula C43H60N2O12. MW 796.9. Isolated from Streptomyces collinus. Antibiotic. Member of the elfermycin group. Antibacterial. Protein biosynthesis inhibitor. Specifically interacts with the bacterial elongation factor EF-Tu and prevents the release of EF-Tu from the bacterial ribosome after GTP hydrolysis. In the absence of ribosomes it induces EF-Tu-dependent hydrolysis of GTP. Animal growth promoter used in poultry breeding.
Catalog No. 50-256-2083
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.