Learn More
Bachem Kisspeptin-13 (4-13) (human) Trifluoroacetate
SDP
Supplier:  Bachem 4037534.0005
Encompass_Preferred
Kisspeptin-13 (4-13) (human) Trifluoroacetate, H-Tyr-Asn-Trp-Asn-Ser-Phe-Gly-Leu-Arg-Phe-NH2, 374675-21-5, C63H83N17O14, Mr 1302.46, 5mg. KiSS-1 (112-121) (human), Malignant Melanoma Metastasis-Suppressor KiSS-1 (112-121) (human), Metastin (45-54) (human), Kisspeptin-14 (5-14) (human), Kisspeptin-54 (45-54) (human), Kisspeptin-10. Kisspeptin-10 (YNWNSFGLRF-amide), an LRF-amide motif-containing fragment of malignant melanoma metastasis suppressor KiSS-1, shows high agonistic potency towards the human G-protein-coupled receptor GPR54 also known as AXOR12. Kisspeptin-10 is a potent stimulator of LH and increases pulse frequency in men.
Catalog No. 50-194-6708
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.