Learn More
MEDPLUS SERVICES USA OtoClear Aquabot Kit (20 OtoClear Tips, 1 Aquabot Bottle, 1 Ear Basin)
Supplier:  MEDPLUS SERVICES USA BNX 7270
Encompass_Preferred
Bionix OtoClear Ear Irrigation helps healthcare workers provide safe and comfortable ear irrigation without the back splash, at half the cost of a self-constructed device. The three-stream tip cleans the ear without directly spraying the tympanic membrane. This is less painful and safer for the patient. Flared tip design prevents over insertion and Threaded design makes this tip compatible with all our delivery devices and any luer lock fittings meeting ISO compliant standards.
Catalog No. NC1858730
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.