Learn More
Cytiva (Formerly Pall Lab) Absorbent pads, non-sterile (1000/pkg)
Supplier:  Cytiva 66190
Encompass
One-Handed Dispensing of Cellulose Absorbent Pads-Enables user to dispense a clean cellulose pad into a Petri dish whenever needed without touching the pad.-Handy dispenser kit holds 1 tube of 100 absorbent pads (10 tubes included). Each tube drops quickly into the hand dispenser for easy use.-Available non-sterile or gammairradiated. No EtO residuals to impede microbial growth. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3292