Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kit Oil no. 10 for Oil Repellency on Paper, Reagents
Kit Oil no. 10 for Oil Repellency on Paper, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Reagents Holdings Llc CK1971001C
Quantity | 1 L |
---|
Chemical Identifiers
108-88-3, 142-82-5 | |
92.14 | |
toluene |
C7H8 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
CC1=CC=CC=C1 |
Specifications
Colorless | |
C7H8 | |
CC1=CC=CC=C1 | |
92.14 | |
Glass Bottle | |
1 L | |
Liquid |
108-88-3, 142-82-5 | |
YXFVVABEGXRONW-UHFFFAOYSA-N | |
toluene | |
Reagent | |
0.7924 | |
Non-sterile | |
Kit Oil #10 for Oil Repellency on Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction