Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kit Oil no. 3 for Oil Repellency on Paper, Reagents

Kit Oil no. 3 for Oil Repellency on Paper, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Supplier:  Reagents Holdings Llc CK1964001C

Quantity 1 L

Catalog No. 01-334-868


Only null left
Add to Cart

Description

Description

Specifications

Chemical Identifiers

8001-79-4 , 108-88-3
933.45
1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
C57H104O9
ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC

Specifications

Colorless
C57H104O9
CCCCCCC(O)CC=CCCCCCCCC(=O)OCC(COC(=O)CCCCCCCC=CCC(O)CCCCCC)OC(=O)CCCCCCCC=CCC(O)CCCCCC
933.45
Glass Bottle
1 L
Liquid
8001-79-4 , 108-88-3
ZEMPKEQAKRGZGQ-UHFFFAOYNA-N
1,3-bis[(12-hydroxyoctadec-9-enoyl)oxy]propan-2-yl 12-hydroxyoctadec-9-enoate
Reagent
0.9298
Non-sterile
Kit Oil #3 for Oil Repellency on Paper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.