Learn More
THERMO SCI LSG 3P Invitrogen™Attune™ NxT Violet 6 Conversion Kit for Yellow Laser
Supplier:  THERMO SCI LSG 3P A36572
Encompass_Preferred
The Attune NxT Violet 6 Conversion kits are designed to reconfigure Attune NxT four-channel violet instruments to allow detection of six channels of violet. A conversion kit is required for each original and any upgraded lasers of the instrument. The conversion is carried out by a Field Service Engineer.
Catalog No. 50-165-6688
Only null left
Add to Cart
Request a Quote
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.