Learn More
ACROBiosystems SARS-CoV-2 Spike protein RBD-Coated Plates, Clear, 96-Well
Supplier:  ACROBiosystems RP135PLATES
Encompass_Preferred
Kits;5PLATES;Streptavidin (SA) has an extraordinarily high affinity for biotin with a dissociation constant (Kd) on the order of 10−14 mol/L, the Biotinylated molecules can bind to the SA irreversibly. The SARS-CoV2 Spike protein RBD-Coated Plates are immobilized with biotinylated SARS-CoV2 Spike protein RBD to a Streptavidin tetramer protein coated plate, which is easy to use and widely available for applications.;Immobilized biotinylated SARS-CoV2 Spike protein RBD at 1 μg/mL (100 μL/well) on Streptavidin Coated Plates, Clear, 96-Well (Cat. No. SP-11), can bind anti-SARS-CoV-2 S1 antibody with a linear range of 0.1-2 ng/mL (QC tested).;Immobilized biotinylated SARS-CoV2 Spike protein RBD at 1 μg/mL (100 μL/well) on Streptavidin Coated Plates, Clear, 96-Well (Cat. No. SP-11), can bind ACE2 with a linear range of 0.2-4 ng/mL (QC tested).
Catalog No. 50-201-9439
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.