Learn More
Thermo Scientific KITTOOLVERTICAL ROTORS
Supplier:  Thermo Scientific 12771
Encompass_Preferred
Centrifuge Accessories; Tool kit; Thermo Scientific; For use with TV-860 vertical titanium rotor and polyallomer 35mL Ultracrimp tubes; Inc. rotor holding fixture, torque wrench, socket; Each
Catalog No. 50-870-293