Learn More
Description
- Environmentally friendly: free of toxic mercury and selenium
- Different tablets to fit individual needs
- For digestion
Specifications
Specifications
| Chemical Name or Material | Titanium |
| Melting Point | 1067°C (Main Component) |
| Color | Blue |
| CAS | 13463-67-7, 7758-99-8, 7778-80-5 |
| Quantity | 1000 Pk. |
| Packaging | Plastic Container |
| Percent Purity | Mixture |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
