Learn More
Microscope World SCHOTT KL 300 LED Cold Light Source
SDP
Supplier:  Microscope World KL300LED
Encompass_Preferred
This SCHOTT LED light source is compact and easy to operate, making it a good choice for use in routine inspections or in education.Color Temperature- 5600KLuminous Flux- Up to 802 lm light flux.Voltage- 100-240V, 50-60Hz.Light Box- Intensity control 0-100%Light Box Dimensions / Weight- 107mm x 114mm x 61mm (4.2in x 4.5in x 2.4in), 0.35kg (0.77 lb)Certifications- CEWarranty- 2 years
Catalog No. NC2965617