Learn More
Cytiva (Formerly Pall Lab) Kleenpak capsule with Emflon II membrane, max diameter 4.2 in, with 1 in. sanitary flange, non-irradiated
Supplier:  Cytiva KA3V002PV1G
Encompass
Kleenpak™ capsule filters with hydrophobic polyvinylidenefluoride (PVDF) Emflon II membrane are especially designed for the sterile filtration of air and gas. These compact filter capsules provide high air flows, resist blocking in wet applications, and provide 100 % removal of bacteria and viruses from air and gases, even in the presence of high humidity and moisture. Kleenpak capsules are manufactured using gamma-irradiation tolerant materials and offer the very highest standard of filtration security and user convenience. These filters can be integrated within gamma-irradiatable or gamma-sterilized complex systems such as biorectors, filter assemblies, or transfer sets where venting is required. Standard pricing not available. Cytiva requires a new quote with updated pricing for each order.
Catalog No. NC0660618
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.