Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark Professional™ Kleenex™ Moisturizing Instant Hand Sanitizer

Catalog No. 19014899 Shop All Kimberly-Clark Professional Products
Change view
Click to view available options
Volume (English):
1 oz.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
19-014-899 1 oz.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-014-899 Supplier Kimberly-Clark Professional™ Supplier No. 36873
This item is not returnable. View return policy

Helps reduce spread of germs at home or business

  • Kills 99.9% of germs in less than 15 seconds
  • Contains moisturizing aloe and vitamin E to soften hands
  • Dermatologist tested and latex compatible

Specifications

Product Type Hand Sanitizer
Composition Moisturizing aloe and vitamin E
Description Latex compatible
Volume (English) 1 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.