Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A20 Breathable Particle Protection Bouffant Cap
GREENER_CHOICE

Catalog No. 19005015 Shop All Ansell Products
Change view
Click to view available options
Size:
One Size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-015 One Size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-005-015 Supplier Ansell Supplier No. 66829
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lightweight and breathable 24 in. bouffant caps are best suited for use in cleanroom applications.

  • Made with breathable Microforce barrier fabric
  • Features serged seams and an elastic opening
  • Elastic is not made with natural rubber latex (reducing risk for individuals with a Type 1 latex allergy)
  • Passes NFPA 99 for antistatic (anti stat) materials
  • Recyclable via The RightCycle Program
TRUSTED_SUSTAINABILITY

Specifications

Product Type Cleanroom Bouffant
Size One Size
Material Rayon
Color White
Disposable Single-use
Model 66829
Seam Type Serged
Certifications/Compliance Passes NFPA 99 for antistatic (anti stat) materials
Packaging Type Cleanroom Packaging
Quantity 500/Cs.
Diameter (English) 24 in.
Diameter (Metric) 60.96 cm
For Use With (Application) Labs, Food Processing, Heavy Dust Environments
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.