Description
- Designed to help protect against dirt, grime and dust
- Five layers of stacked barrier protection (SMMMS) against hazardous particles
- Hip-length shirt
- Comes in a variety of sizes
- Serged seams
- Five snap closures
- Open wrists
- Mandarin collar
- Popular choice for dusty environments such as laboratories and remediation, as well as pesticide applications and as decontamination modesty garments
- Made with breathable Microforce barrier fabric
Specifications
Specifications
| Size | Medium |
| Material | Fabric |
| Color | White |
| Closure Type | Snaps |