Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A40 Reflex™ Liquid and Particle Protection Coveralls
GREENER_CHOICE

Catalog No. 19005128 Shop All Ansell Products
Change view
Click to view available options
Size:
X-Large
2X-Large
3X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-128 3X-Large
19-005-126 X-Large
19-005-127 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-005-128 Supplier Ansell Supplier No. 47999
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Coveralls feature a respirator fit hood, zipper front, elastic wrists and ankles. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals, and dry particulates.

  • Feature respirator fit hood, zipper front, elastic wrists and ankles
  • Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates
  • Pass NFPA 99 for antistatic materials
  • Pass ASTM F1670 testing for penetration of blood and bodily fluids
  • Reflex sizing for a more comfortable fit and less rip outs
  • Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing
TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Coveralls
Size 3X-Large
Gender Unisex
Material Microporous Film Laminate
Color White
Disposable Limited-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type Elastic, Respirator Fit
Closure Type Front Zipper
Seam Type Serged
Sterility Non-sterile
Resistance Liquid, Particle, Anti-static, Chemicals, Bloodborne Pathogens
Certifications/Compliance NFPA 99, ASTM F1670
Model KleenGuard™ A40 Reflex™
No. of Pockets None
For Use With (Application) Aviation, Pharmaceutical Manufacturing, General Manufacturing, Liquid Handling
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.