Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A40 Skid-Resistant Shoe Covers
GREENER_CHOICE

Catalog No. 19005120 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium/Large
Small
X-Large/2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-005-120 Medium/Large
19-005-119 Small
19-005-121 X-Large/2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-005-120 Supplier Ansell Supplier No. 51137
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Helps protect against slips, trips, and falls in a variety of work environments.

  • 7 in. high
  • Made from durable, gray, skid-resistant material
  • Offers up to 74% better traction on wet and dry surfaces than the leading competitor
  • Passes NFPA 99 for antistatic materials
  • Tested to withstand isocyanates and chromate dust
TRUSTED_SUSTAINABILITY

Specifications

Size Medium/Large
Material SMS
Color Gray
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Skid-resistant
Model 51137
For Use With (Application) Painting, Liquid Handling, General Manufacturing, Maintenance
Packaging Quantity 300/Cs.
Height (Metric) 17.78 cm
Product Type Shoe Covers
Show More Show Less

Adhere to
Safety Regulations and Best Practices

Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.