Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ansell KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Lab Coats

Catalog No. 191301581 Shop All Ansell Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Size (English) Chest:
27 in.
28.25 in.
29.75 in.
30 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Size (English) Chest
19-130-1581 2X-Large 30 in.
19-057-440 Medium 27 in.
19-130-1580 X-Large 29.75 in.
19-057-441 Large 28.25 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-1581 Supplier Ansell Supplier No. 45518

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Lab coats offer bloodborne pathogen and chemical splash protection for applications including forensics and animal research

  • Patented three-layer fabric construction
  • Middle layer is a breathable microporous film that offers resistance against many liquids and dry particulates while also allowing heat and sweat vapor to escape
  • Outer layers are a cloth-like yet tough and abrasion-resistant spunbond polypropylene
  • Snap-front, with chest and hip pocket

Compliance

Fabric passes ASTM F1670/F1671 testing for penetration of blood, bodily fluids, and bloodborne pathogens.

TRUSTED_SUSTAINABILITY

Specifications

Product Type General Purpose Lab Coat
Size 2X-Large
Gender Unisex
Material Polypropylene, Microporous Film
Color Blue
Closure Type Snaps
Disposable Single-use
No. of Pockets 2
Seam Type Serged
Size (English) Chest 30 in.
Certifications/Compliance ASTM F1670, F1671
For Use With (Application) Forensics, Animal Research
Construction 3-ply SMS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.