Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KleenGuard™ G40 Latex Coated Cotton Gloves

Catalog No. p-3572462 Shop All Ansell Products
Change view
Click to view available options
Size:
Small
Large
X-Large
2X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-581 Small
19-161-583 Large
19-161-584 X-Large
19-161-585 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Catalog No. 19-161-581 Supplier KleenGuard™ Supplier No. 97270
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
This product contains natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.

Gloves provide a high level of durability in the workplace with a poly-cotton latex coating process to yield lower latex proteins. KleenGuard™ G40 Latex Coated Cotton Gloves ideal for masonry, general maintenance, and warehouse shipping applications.

  • Gloves provide durability in the industrial workplace
  • Poly-cotton latex coating process yields lower latex proteins
  • Crinkle latex texture enhances grip capabilities
  • Breathable cotton back increases comfort
  • Ideal for masonry, construction, and general maintenance
  • Dedicated right and left gloves for a snug, secure fit
TRUSTED_SUSTAINABILITY

Specifications

Coating Material Crinkled Latex
Color Gray
Exterior Finish Fully Textured
Hand Specific
Material Polycotton
Product Type Seamless Knit Gloves
Size Small
Style Full-hand Mitt
For Use With (Application) Masonry, General Maintenance, and Construction
Glove Cuff Color Black
Glove Cuff Style Knit
Series G40
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.