Learn More
Ansell KleenGuard™ Maxfire™ Safety Glasses
Catalog No. 19006204 Shop All Ansell Products
Change view
Click to view available options
:
Clear, anti-fog lens
Catalog No.
19-006-204 Clear, anti-fog lens
1 options
This item is not returnable. View return policy
Catalog No. 19-006-204 Supplier Ansell Supplier No. 38499
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
KleenGuard™ Maxfire Anti-Fog Safety Glasses blend comfort, style, flexibility and protection.
  • Flexible, 5-position, click-to-fit temples and 3-position adjustable nose pad allow users to customize the fit and lens angle for comfort
  • Superior anti-scratch coating passes the EN 166 K mark test for resistance to surface damage by fine particles
  • Anti-fog coating passes the EN 166 N mark
  • Metal-free design with screwless hinges
  • Meets ANSI Z87.1+ standards for high impact
  • Polycarbonate lenses provide 99.9% UVA / UVB / UVC protection
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Impact-resistant, Scratch-resistant
Certifications/Compliance Meets ANSI Z87.1+ standards for high impact (38499-00 meets ANSI Z87.1+2010 and 38499-01 meets ANSI Z87.1+2015)
Lens Tint Clear
Product Type Safety Eyewear
For Use With (Application) General-purpose eye protection
Frame Color Black with red tips
No. per Case 12 pairs/Cs.
Lens Material Polycarbonate
Frame Material Plastic
UV Protection 0.999
Frame Type Black frame with red tips