Learn More
Ansell KleenGuard™ Nemesis™ CSA Safety Glasses
GREENER_CHOICE
Catalog No. 19005074 Shop All Ansell Products
Change view
Click to view available options
Lens Tint:
Amber
Blue, Mirror
Bronze
Green, IR/UV 5.0, Shade 5
Smoke
Frame Color:
Black
Camouflage
Gun Metal
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Lens Tint Frame Color
19-005-074 Green, IR/UV 5.0, Shade 5 Black
19-005-066 Blue, Mirror Black
19-005-070 Bronze Camouflage
19-005-083 Amber Black
19-005-069 Smoke Gun Metal
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-005-074 Supplier Ansell Supplier No. 20640
Add to Cart
Add to Cart
This item is not returnable. View return policy
KleenGuard™ Nemesis™ CSA Safety Glasses feature durable lightweight construction for protection at work and at home.
  • CSA certified and meets ANSI Z87.1+ standards for personal eye protection
  • Sporty, flexible, lightweight design
  • Polycarbonate lenses provide 99.9% UVA/ UVB/UVC protection
  • FLEX-DRY™ nosepiece is designed to be extremely flexible, to channel sweat away from the eyes, and to help prevent slippage down the nose
  • Include several lens options:
    • Bronze lenses provide visual definition with increased contrast and enhanced contours in most light conditions
    • Light blue lenses provide enhanced contrast in sodium vapor lighting environments and bright work environments that use yellow incandescent/fluorescent lamps
    • Copper blue shield lenses have a light brown tint and are designed to filter out blue light for enhanced sharpness and depth perception; ideal for low-light conditions
    • Smoke lenses are worn on bright, sunny days to provide glare reduction; ideal for outdoor use
    • Amber/yellow lenses block out blue light, making the environment appear brighter; ideal for low-light conditions and indoor use
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Impact-resistant, UV-treated
Certifications/Compliance ANSI Z87.1+, CSA Z94.3
Lens Tint Green, IR/UV 5.0, Shade 5
For Use With (Application) Cutting and brazing
Frame Color Black
No. per Case 12 pairs/Cs.
Lens Material Polycarbonate
Frame Material Plastic
UV Protection UV400
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.